siRNA Details
Gene Symbol: C1qa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007572.2 | NP_031598.2 | 3 | 307 | ||
GenBank mRNA | AK002655.1 | - | 3 | 233 | ||
AK136737.1 | - | 3 | 233 | |||
AK150771.1 | - | 3 | 233 | |||
AK159210.1 | - | 3 | 233 | |||
AK159343.1 | - | 3 | 228 | |||
BC002086.1 | - | 3 | 266 | 3592169 | ||
BC020096.1 | - | 2 | 229 | |||
BY318890.1 | - | 3 | 310 | |||
X58861.1 | - | 2 | 230 |
