siRNA Details
Gene Symbol: Atp6v1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007508.5 | NP_031534.2 | 3, 4 | 275 | ||
GenBank mRNA | AK140873.1 | - | 3, 4 | 251 | ||
AK149833.1 | - | 3, 4 | 290 | |||
AK150742.1 | - | 3, 4 | 261 | |||
AK153403.1 | - | 3, 4 | 276 | |||
AK154869.1 | - | 3, 4 | 276 | |||
AK160792.1 | - | 3, 4 | 290 | |||
AK166857.1 | - | 3, 4 | 265 | |||
AK170721.1 | - | 3, 4 | 276 | |||
AK181657.1 | - | 1, 2 | 100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
