siRNA Details
Gene Symbol: NR1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021724.4 | NP_068370.1 | 5 | 1497 | ||
GenBank mRNA | AK315270.1 | - | 6 | 1239 | ||
BC047875.1 | - | 5 | 1321 | 6190140 | ||
BC056148.1 | - | 5 | 1388 | 6195844 | ||
HQ692861.1 | - | 5 | 866 | |||
M24898.1 | - | 5 | 1493 | |||
X72631.1 | - | 5 | 1068 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
