siRNA Details
Gene Symbol: Ciita | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF042158.1 | - | 3 | 390 | ||
AF042159.1 | - | 4 | 696 | |||
AF100709.1 | - | 3 | 573 | |||
AF100710.1 | - | 3 | 289 | |||
AK040723.1 | - | 3 | 452 | |||
AK138822.1 | - | 3 | 482 | |||
AK143956.1 | - | 3 | 561 | |||
AK153609.1 | - | 3 | 386 | |||
AK155184.1 | - | 3 | 413 | |||
AK155311.1 | - | 3 | 482 | |||
BC107330.1 | - | 3 | 331 | |||
BC107331.1 | - | 3 | 331 | |||
U60653.1 | - | 3 | 350 |
