siRNA Details
Gene Symbol: Capn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301250.1 | NP_001288179.1 | 3, 4 | 402 | ||
NM_007602.4 | NP_031628.1 | 3, 4 | 566 | |||
GenBank mRNA | AK155179.1 | - | 3, 4 | 544 | ||
AK164293.1 | - | 3, 4 | 476 | |||
AK170034.1 | - | 3, 4 | 503 | |||
BC014767.1 | - | 3, 4 | 394 | 4458488 | ||
U85020.1 | - | 3, 4 | 485 | |||
Y10656.1 | - | 3, 4 | 498 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_134461, XM_006229729, XM_006229730, XM_006229731, FQ223532, and others.
Gene Ontology Categories:
