siRNA Details
Gene Symbol: Cd59a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111060.2 | NP_001104530.1 | 5 | 785 | ||
NM_007652.5 | NP_031678.1 | 4 | 665 | |||
GenBank mRNA | AK002743.1 | - | 4 | 612 | ||
AK018136.1 | - | 4 | 630 | |||
AK080728.1 | - | 4 | 633 | |||
BC132089.1 | - | 3 | 543 | 40130366 | ||
BC132115.1 | - | 3 | 563 | 40130392 | ||
U60473.1 | - | 4 | 604 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
