siRNA Details
Gene Symbol: Cebpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007679.4 | NP_031705.3 | 1 | 794 | ||
GenBank mRNA | AK149723.1 | - | 259 | |||
AK151958.1 | - | 1 | 795 | |||
BC139293.1 | - | 1 | 794 | 8862315 | ||
X61800.1 | - | 1 | 1070 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
