siRNA Details
Gene Symbol: Psg16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007676.4 | NP_031702.3 | 5 | 1650 | ||
GenBank mRNA | AB500066.1 | - | 4 | 1226 | ||
U34272.1 | - | 5 | 1606 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
