siRNA Details
Gene Symbol: Ephb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BB833527.1 | - | 2 | 447 | ||
BC031924.1 | - | 17 | 3354 | 4911959 | ||
L77867.1 | - | 18 | 3437 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
