siRNA Details
Gene Symbol: Chgb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007694.4 | NP_031720.1 | 4 | 672 | ||
GenBank mRNA | AK019893.1 | - | 4 | 282 | ||
AK077487.1 | - | 4 | 238 | |||
AK077489.1 | - | 4 | 280 | |||
AK133629.1 | - | 4 | 280 | |||
AK133739.1 | - | 4 | 280 | |||
AK133786.1 | - | 4 | 282 | |||
AK158857.1 | - | 4 | 673 | |||
BC014736.1 | - | 4 | 273 | 4511248 | ||
X51429.1 | - | 4 | 268 | |||
X53028.1 | - | 4 | 278 |

More Information
Other Species Targeted

Gene Ontology Categories:
