siRNA Details
Gene Symbol: Socs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168655.1 | NP_001162126.1 | 3 | 635 | ||
NM_001168656.1 | NP_001162127.1 | 3 | 593 | |||
NM_001168657.1 | NP_001162128.1 | 2 | 514 | |||
NM_007706.4 | NP_031732.1 | 3 | 662 | |||
GenBank mRNA | AK033206.1 | - | 1 | 62 | ||
AK137403.1 | - | 3 | 542 | |||
AK144810.1 | - | 3 | 604 | |||
AK160537.1 | - | 3 | 619 | |||
AK162591.1 | - | 3 | 627 | |||
BC106153.1 | - | 3 | 633 | 4989433 | ||
U88327.1 | - | 2 | 422 |
