siRNA Details
Gene Symbol: Cox7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK168077.1 | - | 3 | 353 | ||
BC010772.1 | - | 3 | 347 | 4216583 | ||
X52940.1 | - | 3 | 331 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
