siRNA Details
Gene Symbol: Cryge | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007777.3 | NP_031803.3 | 3 | 315 | ||
GenBank mRNA | AK014301.1 | - | 3 | 302 | ||
BC057012.1 | - | 290 | ||||
BC078417.1 | - | 290 | 6824205 | |||
BC132506.1 | - | 3 | 315 | 40130783 | ||
BC132508.1 | - | 3 | 315 | 40130785 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
