siRNA Details
Gene Symbol: Csn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK085710.1 | - | 4, 5 | 665 | ||
AK142607.1 | - | 4, 5 | 639 | |||
AK164788.1 | - | 4, 5 | 665 | |||
BC049949.1 | - | 4, 5 | 648 | 3963894 | ||
BC050269.1 | - | 4, 5 | 643 | 3966924 | ||
M10114.1 | - | 4, 5 | 623 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
