siRNA Details
Gene Symbol: Csrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK169720.1 | - | 4, 5 | 464 | ||
BC006912.1 | - | 4, 5 | 450 | 2654089 | ||
CT010278.1 | - | 3, 4 | 414 | |||
D88793.1 | - | 4, 5 | 544 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
