siRNA Details
Gene Symbol: Cmah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111110.2 | NP_001104580.1 | 13, 14 | 1905 | ||
NM_001284519.1 | NP_001271448.1 | 14, 15 | 2067 | |||
NM_001284520.1 | NP_001271449.1 | 11, 12 | 1422 | |||
NM_007717.5 | NP_031743.3 | 12, 13 | 1560 | |||
GenBank mRNA | AB061276.1 | - | 13, 14 | 1889 | ||
AB061277.1 | - | 14, 15 | 2051 | |||
AK083394.1 | - | 11, 12 | 1438 | |||
BC028474.1 | - | 13, 14 | 1915 | |||
BC055079.1 | - | 10, 11 | 1419 | 30077348 | ||
D21826.1 | - | 11, 12 | 1532 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
