siRNA Details
Gene Symbol: Arid3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288625.1 | NP_001275554.1 | 2 | 619 | ||
NM_001288626.1 | NP_001275555.1 | 2 | 773 | |||
NM_007880.4 | NP_031906.1 | 2 | 773 | |||
GenBank mRNA | AK141283.1 | - | 2 | 620 | ||
AK154956.1 | - | 2 | 745 | |||
BC050925.1 | - | 2 | 677 | 6831312 | ||
U60335.1 | - | 1 | 561 |
