siRNA Details
Gene Symbol: Epyc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007884.2 | NP_031910.1 | 3 | 246 | ||
GenBank mRNA | AK005684.1 | - | 3 | 247 | ||
AK015223.1 | - | 3 | 246 | |||
AK137179.1 | - | 2 | 171 | |||
AK157900.1 | - | 497 | ||||
AK157901.1 | - | 2 | 508 | |||
BC012695.1 | - | 2 | 218 | 4037028 | ||
D78274.1 | - | 2 | 161 |
