siRNA Details
Gene Symbol: NME4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC017067.1 | - | 4 | 401 | 3913604 | |
BT019438.1 | - | 4 | 370 | |||
CF126980.1 | - | 5 | 553 | |||
Y07604.1 | - | 4 | 381 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
