siRNA Details
Gene Symbol: Usp17la | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007887.2 | NP_031913.1 | 3 | 920 | ||
GenBank mRNA | AY148453.1 | - | 2 | 749 | ||
BC140988.1 | - | 3 | 949 | 9055600 | ||
U41636.1 | - | 3 | 932 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
