siRNA Details
Gene Symbol: Ehf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007914.3 | NP_031940.1 | 9 | 1172 | ||
GenBank mRNA | AF035527.1 | - | 8 | 1170 | ||
AK028651.1 | - | 9 | 1143 | |||
BC005520.1 | - | 10 | 1089 | 3494644 | ||
BC008249.1 | - | 2 | 365 | |||
CF424908.1 | - | 2 | 304 |
