siRNA Details
Gene Symbol: Ddx3x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010028.3 | NP_034158.1 | 17 | 3599 | ||
GenBank mRNA | BC083059.1 | - | 1 | 390 | ||
DQ858457.1 | - | 7 | 2383 | |||
DQ858458.1 | - | 4 | 1924 | |||
DQ858459.1 | - | 3 | 1717 | |||
U42386.1 | - | 1 | 1203 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108246, XM_006256652, XM_006256653, FQ211849, BC085914.
Gene Ontology Categories:
