siRNA Details
Gene Symbol: Ddx3x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010028.3 | NP_034158.1 | 17 | 3790 | ||
GenBank mRNA | BC067210.1 | - | 1 | 573 | ||
BC083059.1 | - | 1 | 581 | |||
DQ858457.1 | - | 7 | 2574 | |||
DQ858458.1 | - | 4 | 2114 | |||
U42386.1 | - | 1 | 1394 |

More Information
Other Species Targeted

Gene Ontology Categories:
