siRNA Details
Gene Symbol: Elk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007922.5 | NP_031948.4 | 4 | 976 | ||
GenBank mRNA | AK132354.1 | - | 4 | 969 | ||
BC054474.1 | - | 4 | 960 | 30045506 | ||
X87257.1 | - | 4 | 1246 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
