siRNA Details
Gene Symbol: Fpr-rs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008041.2 | NP_032067.2 | 1 | 702 | ||
GenBank mRNA | BC115534.1 | - | 1 | 698 | ||
BC115535.1 | - | 1 | 702 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
