siRNA Details
Gene Symbol: Sass6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289568.1 | NP_001276497.1 | 18 | 2987 | ||
NM_001289571.1 | NP_001276500.1 | 16 | 2843 | |||
NM_028349.3 | NP_082625.2 | 17 | 2904 | |||
GenBank mRNA | AK165224.1 | - | 18 | 2935 |
Gene Symbol: Dennd6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134465.2 | NP_001127937.1 | 20 | 5664 | ||
NM_001285466.1 | NP_001272395.1 | 18 | 5510 | |||
NM_001285467.1 | NP_001272396.1 | 20 | 5725 | |||
GenBank mRNA | AK131163.1 | - | 20 | 5636 |
Gene Symbol: Fxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U95736.1 | - | 5 | 955 |
Gene Symbol: Letm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK161638.1 | - | 5 | 2321 |
Gene Symbol: Naa40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089209.1 | - | 1 | 3856 |
Gene Symbol: Psmb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175204.4 | NP_780413.1 | 1 | 2231 | ||
GenBank mRNA | AK037368.1 | - | 1 | 2229 |
Gene Symbol: Usp9x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK053279.1 | - | 254 |
Gene Symbol: Zbtb8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK135238.1 | - | 1 | 2419 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK037888.1 | - | 1405 | Chr. - - [-] on Build |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
