siRNA Details
Gene Symbol: Gm10364 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC099969.1 | - | 2 | 619 |
Gene Symbol: Gm12070 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC098095.1 | - | 2 | 907 |
Gene Symbol: Gm5069 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC064681.1 | - | 2 | 613 |
Gene Symbol: Gm5177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK135747.1 | - | 3 | 1017 |
Gene Symbol: Gm5523 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK188672.1 | - | 138 |
Gene Symbol: Gm6498 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033118.1 | - | 3 | 881 |
Gene Symbol: Gm6981 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132984.1 | - | 3 | 740 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_017008, X02231, BC087743, AB017801, BC087069, and others.
Gene Ontology Categories:
