siRNA Details
Gene Symbol: OR51E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030774.3 | NP_110401.1 | 2 | 795 | ||
GenBank mRNA | AF369708.1 | - | 2 | 701 | ||
AK314051.1 | - | 2 | 795 | |||
AY033942.1 | - | 2 | 792 | |||
BC020768.1 | - | 3 | 668 | 4249775 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
