siRNA Details
Gene Symbol: 3000002C10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013857.1 | - | 1416 | |||
AK082178.1 | - | 1548 | ||||
BC024703.1 | - | 1 | 1118 | |||
BC061210.1 | - | 1 | 1410 |
Gene Symbol: Gm10481 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK160495.1 | - | 437 |
Gene Symbol: Gm12070 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC098095.1 | - | 2 | 1423 |
Gene Symbol: Gm5177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK135747.1 | - | 3 | 1523 |
Gene Symbol: Gm5523 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK190063.1 | - | 120 |
Gene Symbol: Gm6498 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033118.1 | - | 3 | 1394 |
Gene Symbol: Gm6981 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132984.1 | - | 3 | 1255 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK203228.1 | - | 120 | Chr. - - [-] on Build |

More Information
Other Species Targeted

Gene Ontology Categories:
