siRNA Details
Gene Symbol: OR2H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318014.1 | NP_001304943.1 | 4 | 684 | ||
NM_001318022.1 | NP_001304951.1 | 4 | 567 | |||
NM_030883.4 | NP_112145.1 | 4 | 571 | |||
XM_011514470.2 | XP_011512772.1 | 5 | 1968 | |||
XM_011514472.2 | XP_011512774.1 | 4 | 1325 | |||
XM_011514473.2 | XP_011512775.1 | 5 | 1420 | |||
XM_011514479.2 | XP_011512781.1 | 3 | 1161 | |||
XM_011514483.2 | XP_011512785.1 | 3 | 542 | |||
XM_011514484.2 | XP_011512786.1 | 2 | 540 | |||
XM_017010737.1 | XP_016866226.1 | 4 | 2071 | |||
XM_017010738.1 | XP_016866227.1 | 4 | 1452 | |||
XM_017010739.1 | XP_016866228.1 | 4 | 2071 | |||
GenBank mRNA | AJ459847.1 | - | 1 | 76 | ||
AK127984.1 | - | 2 | 785 | |||
BC048991.2 | - | 3 | 519 | 5271584 | ||
DB053988.1 | - | 4 | 556 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
