siRNA Details
Gene Symbol: OR2W1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030903.3 | NP_112165.1 | 1 | 93 | ||
GenBank mRNA | BC103870.1 | - | 1 | 93 | 40004105 | |
BC103871.1 | - | 1 | 93 | 40004106 | ||
BC103872.1 | - | 1 | 93 | 40004109 |
