siRNA Details
Gene Symbol: GPR174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032553.1 | NP_115942.1 | 1 | 141 | ||
GenBank mRNA | AF345567.1 | - | 1 | 141 | ||
BC104922.1 | - | 1 | 141 | 8143925 | ||
BC104924.1 | - | 1 | 141 | 8143927 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
