siRNA Details
Gene Symbol: Gys1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030678.3 | NP_109603.2 | 16 | 2653 | ||
GenBank mRNA | AK050813.1 | - | 14 | 2086 | ||
AK085476.1 | - | 12 | 1681 | |||
AK169344.1 | - | 16 | 2649 | |||
AK170404.1 | - | 16 | 2656 | |||
AK170585.1 | - | 16 | 2576 | |||
AK171148.1 | - | 16 | 2608 | |||
AK171426.1 | - | 16 | 2634 | |||
BC019389.1 | - | 16 | 2452 | 3663506 | ||
X94616.1 | - | 16 | 2501 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
