siRNA Details
Gene Symbol: BRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113182.2 | NP_001106653.1 | 11 | 3530 | ||
NM_001199455.1 | NP_001186384.1 | 11 | 3325 | |||
NM_001199456.1 | NP_001186385.1 | 10 | 1933 | |||
NM_001291986.1 | NP_001278915.1 | 12 | 3622 | |||
NM_005104.3 | NP_005095.1 | 11 | 3617 | |||
GenBank mRNA | AL832722.1 | - | 12 | 3557 | ||
BC063840.1 | - | 11 | 2190 | 6181728 | ||
BX647233.1 | - | 10 | 1933 | |||
BX648109.1 | - | 11 | 3531 | |||
D42040.1 | - | 11 | 3617 | |||
M80613.1 | - | 10 | 2952 | |||
X62083.1 | - | 10 | 2952 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
