siRNA Details
Gene Symbol: TEK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000459.4 | NP_000450.2 | 2 | 634 | ||
NM_001290077.1 | NP_001277006.1 | 2 | 634 | |||
XM_005251561.1 | XP_005251618.1 | 2 | 301 | |||
XM_005251563.1 | XP_005251620.1 | 2 | 301 | |||
GenBank mRNA | AB086825.1 | - | 2 | 290 | ||
AB208796.1 | - | 2 | 312 | |||
AK291775.1 | - | 2 | 322 | |||
AK295043.1 | - | 2 | 360 | |||
BC035514.1 | - | 2 | 309 | 5228999 | ||
DC324016.1 | - | 2 | 561 | |||
EU826591.1 | - | 2 | 184 | |||
EU826592.1 | - | 2 | 184 | |||
L06139.1 | - | 2 | 332 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
