siRNA Details
Gene Symbol: Gstm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008184.3 | NP_032210.3 | 8 | 699 | ||
GenBank mRNA | AJ000413.1 | - | 8 | 583 | ||
BC031818.1 | - | 8 | 662 | 4196506 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
