siRNA Details
Gene Symbol: Gys1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030678.3 | NP_109603.2 | 16 | 2713 | ||
GenBank mRNA | AK050813.1 | - | 14 | 2146 | ||
AK085476.1 | - | 12 | 1741 | |||
AK169344.1 | - | 16 | 2709 | |||
AK170404.1 | - | 16 | 2716 | |||
AK170585.1 | - | 16 | 2636 | |||
AK171148.1 | - | 16 | 2668 | |||
AK171426.1 | - | 16 | 2694 | |||
X94616.1 | - | 16 | 2561 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
