siRNA Details
Gene Symbol: Hgs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159328.1 | NP_001152800.1 | 3 | 256 | ||
NM_008244.3 | NP_032270.3 | 3 | 256 | |||
GenBank mRNA | AK046299.1 | - | 3 | 217 | ||
AK145031.1 | - | 3 | 176 | |||
AK166441.1 | - | 3 | 230 | |||
AK195076.1 | - | 3 | 150 | |||
BC003239.1 | - | 3 | 200 | 3497966 | ||
D50050.1 | - | 3 | 206 |
