siRNA Details
Gene Symbol: Hsd17b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008292.4 | NP_032318.2 | 2, 3 | 256 | ||
GenBank mRNA | AK004866.1 | - | 2, 3 | 256 | ||
AK088381.1 | - | 2, 3 | 231 | |||
AK147182.1 | - | 2, 3 | 228 | |||
AK152532.1 | - | 2, 3 | 151 | |||
AK161655.1 | - | 2, 3 | 151 | |||
AK166801.1 | - | 2, 3 | 169 | |||
AK167060.1 | - | 2, 3 | 206 | |||
AK169077.1 | - | 2, 3 | 158 | |||
AK205578.1 | - | 1, 2 | 55 | |||
X89998.1 | - | 2, 3 | 117 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
