siRNA Details
Gene Symbol: Hsf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008297.3 | NP_032323.3 | 2 | 270 | ||
GenBank mRNA | AK053976.1 | - | 2 | 271 | ||
BC018414.1 | - | 2 | 192 | 3990765 | ||
X61754.1 | - | 2 | 304 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
