siRNA Details
Gene Symbol: Hspg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008305.3 | NP_032331.2 | 2 | 175 | ||
GenBank mRNA | AK147294.1 | - | 2 | 176 | ||
AK147396.1 | - | 2 | 176 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
