siRNA Details
Gene Symbol: MCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197320.1 | NP_001184249.1 | 2, 3 | 445 | ||
NM_021960.4 | NP_068779.1 | 1, 2 | 904 | |||
GenBank mRNA | AF118124.1 | - | 1, 2 | 745 | ||
AF118276.1 | - | 1, 2 | 154 | |||
AF118277.1 | - | 1, 2 | 154 | |||
AF118278.1 | - | 1, 2 | 154 | |||
AK294462.1 | - | 2, 3 | 476 | |||
AK297217.1 | - | 2, 3 | 440 | |||
AK300499.1 | - | 2, 3 | 550 | |||
AK304775.1 | - | 2, 3 | 586 | |||
AK312508.1 | - | 1, 2 | 776 | |||
AK316267.1 | - | 2, 3 | 732 | |||
BC017197.2 | - | 1, 2 | 750 | 3138465 | ||
BC071897.1 | - | 1, 2 | 752 | 6586159 | ||
BC107735.1 | - | 1, 2 | 767 | 4770487 | ||
BT006640.1 | - | 1, 2 | 696 | |||
L08246.1 | - | 1, 2 | 756 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
