siRNA Details
Gene Symbol: XYLB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005108.3 | NP_005099.2 | 3 | 285 | ||
XM_011534325.2 | XP_011532627.1 | 3 | 266 | |||
XM_011534327.1 | XP_011532629.1 | 3 | 266 | |||
XM_011534328.2 | XP_011532630.1 | 3 | 266 | |||
XM_011534329.1 | XP_011532631.1 | 3 | 266 | |||
XM_011534330.2 | XP_011532632.1 | 3 | 266 | |||
XM_017007596.1 | XP_016863085.1 | 3 | 311 | |||
XM_017007598.1 | XP_016863087.1 | 3 | 266 | |||
XM_017007599.1 | XP_016863088.1 | 3 | 266 | |||
GenBank mRNA | AB015046.1 | - | 3 | 305 | ||
AK025728.1 | - | 3 | 237 | |||
AK314386.1 | - | 3 | 233 | |||
BC030231.2 | - | 3 | 254 | |||
BC137076.1 | - | 3 | 217 | 9021073 | ||
BC137080.1 | - | 3 | 217 | 9021077 | ||
BI090475.1 | - | 3 | 148 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
