siRNA Details
Gene Symbol: Hyal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008317.4 | NP_032343.2 | 4 | 2063 | ||
GenBank mRNA | AF011567.1 | - | 4 | 2051 | ||
AF417496.1 | - | 1008 | ||||
AF417497.1 | - | 1008 | ||||
AF417498.1 | - | 1008 | ||||
BC021636.1 | - | 4 | 2064 | 3990455 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
