siRNA Details
Gene Symbol: Irgm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008326.1 | NP_032352.1 | 3 | 1819 | ||
GenBank mRNA | AK002545.1 | - | 3 | 1841 | ||
AK167558.1 | - | 2 | 1673 | |||
AK171743.1 | - | 3 | 1800 | |||
BC145957.1 | - | 3 | 1631 | 40131260 | ||
U19119.1 | - | 3 | 1819 |
