siRNA Details
Gene Symbol: Cep250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129999.1 | NP_001123471.1 | 28 | 6183 | ||
NM_001130000.1 | NP_001123472.1 | 28 | 6181 | |||
NM_008383.3 | NP_032409.3 | 28 | 6123 | |||
NM_177217.3 | NP_796191.2 | 28 | 6118 | |||
GenBank mRNA | AK139187.1 | - | 1 | 23 | ||
BC068153.1 | - | 26 | 5360 | |||
BC075730.1 | - | 1 | 809 | |||
BC086623.1 | - | 10 | 4565 | |||
BC145268.1 | - | 25 | 5763 | |||
BC150746.1 | - | 27 | 5945 | 9087657 | ||
DQ148475.1 | - | 27 | 5918 | |||
U33198.1 | - | 1 | 501 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
