siRNA Details
Gene Symbol: XYLB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005108.3 | NP_005099.2 | 19 | 3078 | ||
GenBank mRNA | AK022601.1 | - | 2 | 1535 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
