siRNA Details
Gene Symbol: Gimap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008376.3 | NP_032402.2 | 3 | 1320 | ||
NM_175860.2 | NP_787056.1 | 3 | 1314 | |||
GenBank mRNA | AK156732.1 | - | 3 | 1315 | ||
AK169133.1 | - | 3 | 1273 | |||
BC132601.1 | - | 3 | 1280 | 40130878 | ||
BC132603.1 | - | 3 | 1281 | 40130880 | ||
Y08026.2 | - | 1 | 1429 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
