siRNA Details
Gene Symbol: Tm4sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008536.3 | NP_032562.1 | 5 | 961 | ||
GenBank mRNA | AK077326.1 | - | 3 | 464 | ||
AK133556.1 | - | 462 | ||||
AK134603.1 | - | 4 | 897 | |||
AK164718.1 | - | 3 | 465 | |||
BC083073.1 | - | 5 | 961 | 6337376 | ||
KC508089.1 | - | 3 | 436 | |||
L15429.1 | - | 3 | 451 | |||
L15443.1 | - | 3 | 451 |
