siRNA Details
Gene Symbol: Klra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF253057.1 | - | 4 | 530 | ||
AF253058.1 | - | 4 | 530 | |||
AF253059.1 | - | 4 | 530 | |||
AY860971.1 | - | 4 | 530 | |||
U10304.1 | - | 5 | 624 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
